Triisopropoxyvanadium(V) oxide ≥98%

Supplier: STREM CHEMICALS INC MS
Warning

Synonyms: Vanadium(V) Triisopropoxide Oxide, Vanadium (V) tri-i-propoxy oxide, Vanadium(V) Oxytriisopropoxide, VTIP

23-5000-5G 23-5000-100G 23-5000-25G
100201-536EA 80.68 USD
100201-536 100206-488 100201-534
Triisopropoxyvanadium(V) oxide ≥98%
Triisopropoxyvanadium(V) oxide

Metal Alkoxides, Volatile Precursors for CVD

Formula: C₉H₂₁O₄V
MW: 244.20 g/mol
Boiling Pt: 60…61 °C (0.5 mmHg)
Density: 1,035
Flash Pt: 113 °F
MDL Number: MFCD00015017
CAS Number: 5588-84-1
UN: 1993
ADR: 3,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR