Vanadium(III) chloride tetrahydrofuran complex (1:3)

Supplier: STREM CHEMICALS INC MS
Danger

Synonyms: Trichlorotris(tetrahydrofuran)vanadium, Vanadium(III) chloride tetrahydrofuran adduct, Trichlorotris(tetrahydrofuran)vanadium(III)

23-4350-5G 23-4350-25G
100201-532EA 100.49 USD
100201-532 100208-624
Vanadium(III) chloride tetrahydrofuran complex (1:3)
Vanadium(III) chloride tetrahydrofuran complex (1:3)
Formula: VCl₃·3C₄H₈O
MW: 373.62 g/mol
MDL Number: MFCD00145515
CAS Number: 19559-06-9
UN: 1759
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR