Vanadyl(IV) acetylacetonate ≥98%

Supplier: STREM CHEMICALS INC MS
Warning

Synonyms: Vanadium(IV) oxide acetylacetonate, Oxobis(2,4-pentanedionato-O,O')vanadium (IV), Vanadium(IV) bis(acetylacetonato)oxide, Vanadyl acetylacetonate, Di(2,4-pentandionato-O,O')vanadium(IV)oxid, Vanadium (IV) oxide bis(2,4-pentanedionate)

23-2202-50G 23-2202-250G
100206-486EA 99.08 USD
100206-486 100202-374
Vanadyl(IV) acetylacetonate ≥98%
Vanadyl(IV) acetylacetonate
Formula: C₁₀H₁₄O₅V
MW: 265.16 g/mol
Melting Pt: 250 °C (dec.)
MDL Number: MFCD00000032
CAS Number: 3153-26-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR