Trichlorotris(tetrahydrofuran)titanium(III) ≥97%
Supplier: STREM CHEMICALS INC MS
|
Danger
|
Synonyms:
Titanium(III) chloride tetrahydrofuran complex
22-3500-25G
22-3500-5G
22-3500-100G
100208-382EA
748.74
USD
100208-382
100208-384
100202-490
Trichlorotris(tetrahydrofuran)titanium(III) ≥97%
Trichlorotris(tetrahydrofuran)titanium(III)
Formula:
C₁₂H₂₄Cl₃O₃Ti MW: 370.543 g/mol |
MDL Number:
MFCD01631276 CAS Number: 18039-90-2 UN: 1325 ADR: 4.1,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...