Titanium(III) chloride aluminum(III) chloride ≥98%

Supplier: STREM CHEMICALS INC MS
Danger

Synonyms: Titanium(III) chloride, Al reduced, Titanium aluminum chloride, Titanium(III) chloride, Aluminum titanium chloride

22-1180-500G 22-1180-100G
100203-044EA 784.12 USD
100203-044 100208-380
Titanium(III) chloride aluminum(III) chloride ≥98%
Titanium(III) chloride aluminum(III) chloride
Formula: (TiCl₃)₃AlCl₃
MW: 596.15 g/mol
MDL Number: MFCD00145412
CAS Number: 12003-13-3
UN: 2869
ADR: 8 II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR