(Pentamethylcyclopentadienyl)titanium trichloride ≥98%
Supplier: STREM CHEMICALS INC MS
|
Danger
|
Synonyms:
Trichloro(pentamethylcyclopentadienyl)titanium(IV)
22-1075-5G
22-1075-1G
100206-472EA
441.6
USD
100206-472
100206-470
(Pentamethylcyclopentadienyl)titanium trichloride ≥98%
(Pentamethylcyclopentadienyl)titanium trichloride
Metallocenes, Derivatives & Cp Precursors
Formula:
C₁₀H₁₅Cl₃Ti MW: 289.45 g/mol Melting Pt: 225 °C |
MDL Number:
MFCD00070434 CAS Number: 12129-06-5 UN: 1759 ADR: 8 II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...