Tetrakis(diethylamino)titanium (TDEAT) ≥99%

Supplier: STREM CHEMICALS INC MS
Danger

Synonyms: Tetrakis(diethylamino)titanium(IV)

22-1050-5G 22-1050-25G
100196-144EA 130.22 USD
100196-144 100196-142
Tetrakis(diethylamino)titanium (TDEAT) ≥99%
Tetrakis(diethylamino)titanium (TDEAT)

Metal Alkylamides, Volatile Precursors for CVD

Formula: C₁₆H₄₀N₄Ti
MW: 336.383 g/mol
Boiling Pt: 133 °C (1.2 mmHg)
Density: 0.938
Flash Pt: –18 °F
MDL Number: MFCD00015648
CAS Number: 4419-47-0
UN: 3399
ADR: 4.3,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR