Hydrotris(pyrazol-1-ylborato)trichlorotitanium(IV) ≥95%

Supplier: STREM CHEMICALS INC MS
Danger

22-0900-1G 22-0900-250MG
100209-330EA 421.78 USD
100209-330 100199-824
Hydrotris(pyrazol-1-ylborato)trichlorotitanium(IV) ≥95%
Hydrotris(pyrazol-1-ylborato)trichlorotitanium(IV)
MDL Number: MFCD00269848
CAS Number: 58097-69-1
UN: 1759
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR