Bis(2,2,6,6-tetramethyl-3,5-heptanedionato-O,O')titanium(IV) dichloride ≥99%

Supplier: STREM CHEMICALS INC MS
Warning

22-0600-25G 22-0600-1G 22-0600-5G
100204-532EA 853.48 USD
100204-532 100196-140 100209-772
Bis(2,2,6,6-tetramethyl-3,5-heptanedionato-O,O')titanium(IV) dichloride ≥99%
Bis(2,2,6,6-tetramethyl-3,5-heptanedionato-O,O')titanium(IV) dichloride

Metal TMHD

Formula: C₂₂H₃₈Cl₂O₄Ti
Boiling Pt: 215 °C (dec.)
Melting Pt: 127…129 °C
MDL Number: MFCD00144106
CAS Number: 53293-32-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR