Bis(2,2,6,6-tetramethyl-3,5-heptanedionato-O,O')titanium(IV) oxide ≥95%

Supplier: STREM CHEMICALS INC MS

Synonyms: OTi(TMHD)2, Bis(2,2,6,6-tetramethyl-3,5-heptanedionato)oxotitanium(IV)

22-0340-500MG 22-0340-2G 22-0340-10G
100203-756EA 191.08 USD
100203-756 100207-618 100198-154
Bis(2,2,6,6-tetramethyl-3,5-heptanedionato-O,O')titanium(IV) oxide ≥95%
Bis(2,2,6,6-tetramethyl-3,5-heptanedionato-O,O')titanium(IV) oxide

Metal TMHD

Formula: C₂₂H₃₈O₅Ti
MW: 430.4 g/mol
Melting Pt: 120 °C (dec.)
MDL Number: MFCD00269847
CAS Number: 152248-67-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR