Bis(i-propylcyclopentadienyl)titanium dichloride ≥98%

Supplier: STREM CHEMICALS INC MS
Warning

22-0320-5G 22-0320-1G
100202-372EA 707.69 USD
100202-372 100205-008
Bis(i-propylcyclopentadienyl)titanium dichloride ≥98%
Bis(i-propylcyclopentadienyl)titanium dichloride

Metallocenes, Derivatives & Cp Precursors

MDL Number: MFCD01862448
CAS Number: 12130-65-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR