Bis(pentamethylcyclopentadienyl)titanium(IV) dichloride ≥99%

Supplier: STREM CHEMICALS INC MS
Warning

Synonyms: Bis(pentamethylcyclopentadienyl)titanium dichloride

22-0300-1G 22-0300-5G
100205-004EA 188.25 USD
100205-004 100205-006
Bis(pentamethylcyclopentadienyl)titanium(IV) dichloride ≥99%
Bis(pentamethylcyclopentadienyl)titanium(IV) dichloride

Metallocenes, Derivatives & Cp Precursors

Formula: C₂₀H₃₀Cl₂Ti
MW: 389.225 g/mol
MDL Number: MFCD00049153
CAS Number: 11136-36-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR