Bis(ethylcyclopentadienyl)titanium(IV) dichloride ≥98%

Supplier: STREM CHEMICALS INC MS
Warning

22-0250-5G 22-0250-1G
100202-370EA 728.92 USD
100202-370 100205-002
Bis(ethylcyclopentadienyl)titanium(IV) dichloride ≥98%
Bis(ethylcyclopentadienyl)titanium(IV) dichloride

Metallocenes, Derivatives & Cp Precursors

Formula: C₁₄H₁₈Cl₂Ti
MDL Number: MFCD01862447
CAS Number: 35625-75-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR