Dicarbonylbis(cyclopentadienyl)titanium(II) ≥98%
Supplier: STREM CHEMICALS INC MS
|
Danger
|
Synonyms:
Bis(cyclopentadienyl)dicarbonyl titanium(II)
22-0180-1G
22-0180-5G
100205-832EA
213.72
USD
100205-832
100205-834
Dicarbonylbis(cyclopentadienyl)titanium(II) ≥98%
Dicarbonylbis(cyclopentadienyl)titanium(II)
Metallocenes, Derivatives & Cp Precursors
Formula:
C₁₂H₁₀O₂Ti MW: 234.076 g/mol |
MDL Number:
MFCD00143762 CAS Number: 12129-51-0 UN: 1325 ADR: 4.1,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...