Bis(tert-butylcyclopentadienyl)titanium(IV) dichloride ≥98%

Supplier: STREM CHEMICALS INC MS
Warning

Synonyms: Bis(t-butylcyclopentadienyl)titanium dichloride

22-0170-1G 22-0170-25G 22-0170-5G
100198-150EA 205.23 USD
100198-150 100207-368 100206-098
Bis(tert-butylcyclopentadienyl)titanium(IV) dichloride ≥98%
Bis(tert-butylcyclopentadienyl)titanium(IV) dichloride

Metallocenes, Derivatives & Cp Precursors

Formula: C₁₈H₂₆Cl₂Ti
MW: 361.172 g/mol
MDL Number: MFCD01073774
CAS Number: 79269-71-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR