Bis(6,6,7,7,8,8,8-heptafluoro-2,2-dimethyl-3,5-octanedionate)calcium

Supplier: STREM CHEMICALS INC MS

Synonyms: Ca(FOD)2

20-8400-25G 20-8400-1G 20-8400-5G
100206-096EA 1309.23 USD
100206-096 100206-462 100197-096
Bis(6,6,7,7,8,8,8-heptafluoro-2,2-dimethyl-3,5-octanedionate)calcium
Bis(6,6,7,7,8,8,8-heptafluoro-2,2-dimethyl-3,5-octanedionate)calcium

Metal Beta-diketonates, Volatile Precursors for CVD

Boiling Pt: 250 °C/ 170 °C (0.1 mmHg) (dec./ subl.)
Melting Pt: 208…210 °C
MDL Number: MFCD00070431
CAS Number: 36885-29-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR