Calcium neodecanoate (9 - 11% Ca), Superconductor Grade

Supplier: STREM CHEMICALS INC MS

20-7000-50G 20-7000-10G
100199-366EA 489.72 USD
100199-366 100205-000
Calcium neodecanoate (9 - 11% Ca), Superconductor Grade
Calcium neodecanoate
MDL Number: MFCD00070430
CAS Number: 27253-33-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR