Calcium trifluoromethanesulfonate ≥96%

Supplier: STREM CHEMICALS INC MS
Warning

Synonyms: Calcium trifluoromethanesulphonate, Calcium triflate

20-5000-10G 20-5000-50G
100199-822EA 140.12 USD
100199-822 100202-646
Calcium trifluoromethanesulfonate ≥96%
Calcium trifluoromethanesulphonate

Metal Triflate

Formula: C₂CaF₆O₆S₂
MW: 338.22 g/mol
Melting Pt: Dec.
MDL Number: MFCD00143911
CAS Number: 55120-75-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR