Calcium hexafluoroacetylacetonate dihydrate ≥97%

Supplier: STREM CHEMICALS INC MS
Warning

20-2500-5G 20-2500-25G 20-2500-1G
100204-530EA 281.66 USD
100204-530 100209-770 100198-696
Calcium hexafluoroacetylacetonate dihydrate ≥97%
Calcium hexafluoroacetylacetonate dihydrate

Metal Beta-diketonates, Volatile Precursors for CVD

Boiling Pt: 230…240 °C/ 180 °C (0.07 mmHg) (dec./ subl.)
Melting Pt: 135…140 °C
MDL Number: MFCD00151016
CAS Number: 203863-17-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR