Calcium chloride hydrate ≥99.999% (Ca basis), PURATREM

Supplier: STREM CHEMICALS INC MS
Warning

20-2060-100G 20-2060-25G 20-2060-5G
100200-438EA 1261.11 USD
100200-438 100206-094 100206-460
Calcium chloride hydrate ≥99.999% (Ca basis), PURATREM
Calcium chloride hydrate
Formula: CaCl₂·H₂O
MW: 110.98 g/mol (anhy)
MDL Number: MFCD00149613
CAS Number: 22691-02-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR