Calcium wolframate ≥98%
Supplier: STREM CHEMICALS INC MS
Synonyms:
Calcium tungstate, Calcium tungsten oxide
20-2052-100G
20-2052-500G
100204-998EA
82.09
USD
100204-998
100200-258
Calcium wolframate ≥98%
Calcium wolframate
Formula:
CaO₄W MW: 287.92 g/mol Melting Pt: 1620 °C Density: 6,062 |
MDL Number:
MFCD00010913 CAS Number: 7790-75-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...