Calcium bis(dihydrogenorthophosphate) monohydrate ≥99%

Supplier: STREM CHEMICALS INC MS

20-2037-1000G 20-2037-250G
100204-008EA 257.6 USD
100204-008 100199-820
Calcium bis(dihydrogenorthophosphate) monohydrate ≥99%
Calcium bis(dihydrogenorthophosphate) monohydrate
Formula: H₄CaO₈P₂·1H₂O
MW: 252.07 g/mol
MDL Number: MFCD00149602
CAS Number: 10031-30-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR