Bis(2,2,6,6-tetramethyl-3,5-heptanedionato)calcium ≥97%

Supplier: STREM CHEMICALS INC MS

Synonyms: Ca(TMHD)2

20-1000-5G 20-1000-1G
100201-514EA 297.23 USD
100201-514 100201-510 100201-512
Bis(2,2,6,6-tetramethyl-3,5-heptanedionato)calcium ≥97%
Bis(2,2,6,6-tetramethyl-3,5-heptanedionato)calcium

Metal Beta-diketonates, Metal TMHD, Volatile Precursors for CVD

Formula: C₂₂H₃₈CaO₄
MW: 406.57 g/mol
Boiling Pt: 280 °C/ 205 °C (0,1 mmHg) (dec./ subl.)
Melting Pt: 220…223 °C
MDL Number: MFCD00064757
CAS Number: 118448-18-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR