Potassium trioxalatoferrate(III) trihydrate

Supplier: STREM CHEMICALS INC MS

19-5000-10G 19-5000-50G
100197-438EA 69.35 USD
100197-438 100196-132
Potassium trioxalatoferrate(III) trihydrate
Potassium trioxalatoferrate(III) trihydrate
Formula: K₃Fe(C₂O₄)₃·3H₂O
MDL Number: MFCD00150450
CAS Number: 5936-11-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR