Potassium hydrotris(3-(6-methyl-3-pyridyl)-5-methylpyrazol-1-yl)borate ≥97%

Supplier: STREM CHEMICALS INC MS
Warning

19-3400-250MG 19-3400-1G
100208-370EA 140.12 USD
100208-370 100198-692
Potassium hydrotris(3-(6-methyl-3-pyridyl)-5-methylpyrazol-1-yl)borate ≥97%
Potassium hydrotris(3-(6-methyl-3-pyridyl)-5-methylpyrazol-1-yl)borate
Melting Pt: 198 °C
MDL Number: MFCD01862444
CAS Number: 184032-07-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR