Potassium hydrogen sulfide, anhydrous ≥95%

Supplier: STREM CHEMICALS INC MS
Danger

19-3300-1G 19-3300-5G
101442-450EA 140.12 USD
101442-450 101442-452
Potassium hydrogen sulfide, anhydrous ≥95%
Potassium hydrogen sulfide
MDL Number: MFCD00061365
CAS Number: 1310-61-8
UN: 3190
ADR: 4.2 II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR