Potassium tris(3,5-dimethylpyrazol-1-yl)borohydride ≥97%

Supplier: STREM CHEMICALS INC MS
Warning

Synonyms: Potassium Hydrotris(3,5-dimethylpyrazol-1-yl)borate, Potassium hydrotris(3,5-dimethylpyrazol-1-yl)borate

19-2900-1G 19-2900-5G
100207-608EA 120.31 USD
100207-608 100197-094
Potassium tris(3,5-dimethylpyrazol-1-yl)borohydride ≥97%
Potassium tris(3,5-dimethylpyrazol-1-yl)borohydride
Formula: C₁₅H₂₂BKN₆
MW: 336.29 g/mol
Melting Pt: 298…300 °C
MDL Number: MFCD00040342
CAS Number: 17567-17-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR