Potassium tri-sec-butylborohydride 1.0M in THF
Supplier: STREM CHEMICALS INC MS
|
Danger
|
Packed in a Sure/Seal™ bottle (Trademark of Sigma-Aldrich Co. LLC).
19-1965-500MMOL
19-1965-100MMOL
100200-252EA
1568.25
USD
100200-252
100205-830
Potassium tri-sec-butylborohydride 1.0M in THF
Potassium tri-sec-butylborohydride
Formula:
C₁₂H₂₇BK Flash Pt: 1 °F |
MDL Number:
MFCD00009324 CAS Number: 54575-49-4 UN: 3399 ADR: 4.3,I |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...