Potassium diphenylbis(pyrazol-1-yl)borate ≥98%

Supplier: STREM CHEMICALS INC MS
Warning

19-1700-1G 19-1700-250MG
100209-476EA 421.78 USD
100209-476 100203-018
Potassium diphenylbis(pyrazol-1-yl)borate ≥98%
Potassium diphenylbis(pyrazol-1-yl)borate
Melting Pt: 283…285 °C
MDL Number: MFCD02684554
CAS Number: 109088-11-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR