(R)-(+)-2-Methyl-2-propanesulfinamide ≥97%

Supplier: STREM CHEMICALS INC MS
Warning

Synonyms: (R)-2-methylpropane-2-sulphinamide, R)-(+)-tert-Butyl sulphinamide, (R)-2-Methyl-2-propanesulphinamide, (R)-(+)-tert-Butylsulphinamide, (R)-(+)-t-Butylsulphinamide

16-0380-1G 16-0380-5G
101324-638EA 169.85 USD
101324-638 101324-640
(R)-(+)-2-Methyl-2-propanesulfinamide ≥97%
(R)-(+)-2-Methyl-2-propanesulfinamide
Formula: C₄H₁₁NOS
MW: 121.20 g/mol
Melting Pt: 103…107 °C
MDL Number: MFCD05861479
CAS Number: 196929-78-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR