Bis(dicyclohexylphosphino)methane ≥97%

Supplier: STREM CHEMICALS INC MS
Warning

15-9590-5G 15-9590-1G
100206-448EA 694.95 USD
100206-448 100206-446
Bis(dicyclohexylphosphino)methane ≥97%
Bis(dicyclohexylphosphino)methane

Phosphine

Formula: C₂₅H₄₆P₂
MDL Number: MFCD00143763
CAS Number: 137349-65-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR