1,2-Bis(dicyclohexylphosphino)ethane ≥98%

Supplier: STREM CHEMICALS INC MS
Warning

15-9585-1G 15-9585-5G
100208-364EA 140.12 USD
100208-364 100208-366
1,2-Bis(dicyclohexylphosphino)ethane ≥98%
1,2-Bis(dicyclohexylphosphino)ethane

Phosphine

Formula: C₂₆H₄₈P₂
MW: 422.607 g/mol
Melting Pt: 96…97 °C
MDL Number: MFCD00015521
CAS Number: 23743-26-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR