2,2'-Bis(dicyclohexylphosphino)-1,1'-biphenyl ≥97%

Supplier: STREM CHEMICALS INC MS
Warning

15-9560-250MG 15-9560-1G
101324-630EA 89.17 USD
101324-630 101324-632
2,2'-Bis(dicyclohexylphosphino)-1,1'-biphenyl ≥97%
2,2'-Bis(dicyclohexylphosphino)-1,1'-biphenyl

Phosphine

Formula: C₃₆H₅₂P₂
MW: 546.7456 g/mol
MDL Number: MFCD07368358
CAS Number: 255897-36-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR