Vinyldiphenylphosphine ≥97%

Supplier: STREM CHEMICALS INC MS

Synonyms: Diphenyl(vinyl)phosphine

15-9150-25G 15-9150-5G 15-9150-1G
100207-238EA 702.03 USD
100207-238 100203-746 100197-432
Vinyldiphenylphosphine ≥97%
Vinyldiphenylphosphine

Phosphine

Formula: C₁₄H₁₃P
MW: 212.2268 g/mol
Boiling Pt: 135 °C (3,5 mmHg)
Density: 1,067
Flash Pt: >230 °F
MDL Number: MFCD00008611
CAS Number: 2155-96-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR