Tris(trimethylsilyl)phosphine ≥98%

Supplier: STREM CHEMICALS INC MS
Danger

15-8020-1G 15-8020-5G
100197-090EA 178.34 USD
100197-090 100197-092
Tris(trimethylsilyl)phosphine ≥98%
Tris(trimethylsilyl)phosphine

Phosphine

Formula: [(CH₃)₃Si]₃P
Boiling Pt: 102…105 °C (16 mmHg)
Melting Pt: 24 °C
Density: 0,863
Flash Pt: –1 °F
MDL Number: MFCD00015487
CAS Number: 15573-38-3
UN: 2845
ADR: 4.2,I

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR