Triphenylphosphine-3,3',3''-trisulfonic acid trisodium salt hydrate (<10% oxide)

Supplier: STREM CHEMICALS INC MS
Warning

Synonyms: Trisodium 3,3',3''-Phosphinetriyltribenzenesulphonate hydrate, Sodium 3,3',3''-phosphinetriyltribenzenesulphonate hydrate, Tris(3-sulphophenyl)phosphinetrisodium salt hydrate, Tris(3-sulphonatophenyl)phosphine hydrate, sodium salt hydrate

15-8007-5G 15-8007-1G
100200-872EA 1592.31 USD
100200-872 100196-126
Triphenylphosphine-3,3',3''-trisulfonic acid trisodium salt hydrate (<10% oxide)
Triphenylphosphine-3,3',3''-trisulfonic acid trisodium salt hydrate

Phosphine, Phosphine, Water Soluble

Formula: C₁₈H₁₂Na₃O₉PS₃.xH₂O
MDL Number: MFCD11046357
CAS Number: 63995-70-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR