Tris(4-methoxy-3,5-dimethylphenyl)phosphine ≥98%
Supplier: STREM CHEMICALS INC MS
|
Warning
|
15-7940-5G
15-7940-1G
100199-814EA
543.51
USD
100199-814
100199-812
Tris(4-methoxy-3,5-dimethylphenyl)phosphine ≥98%
Tris(4-methoxy-3,5-dimethylphenyl)phosphine
Phosphine
Formula:
C₂₇H₃₃O₃P MW: 436.52 g/mol Melting Pt: 175…177 °C |
MDL Number:
MFCD03840342 CAS Number: 121898-64-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...