Tris(hydroxymethyl)phosphine ≥85%

Supplier: STREM CHEMICALS INC MS
Danger

15-7900-1G 15-7900-5G
100209-746EA 113.23 USD
100209-746 100196-122
Tris(hydroxymethyl)phosphine ≥85%
Tris(hydroxymethyl)phosphine

Phosphine, Phosphine, Water Soluble

Formula: C₃H₉O₃P
MW: 124.076 g/mol
MDL Number: MFCD00055382
CAS Number: 2767-80-8
UN: 2811
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR