1,1,1-Tris(diphenylphosphino)methane ≥97%

Supplier: STREM CHEMICALS INC MS
Warning

Synonyms: Tris(diphenylphosphino)methane

15-7880-1G 15-7880-5G
100207-598EA 134.46 USD
100207-598 100203-016
1,1,1-Tris(diphenylphosphino)methane ≥97%
1,1,1-Tris(diphenylphosphino)methane

Phosphine

Formula: C₃₇H₃₁P₃
MW: 568.563 g/mol
Melting Pt: 168…172 °C
MDL Number: MFCD00015527
CAS Number: 28926-65-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR