Triphos ≥97%

Supplier: STREM CHEMICALS INC MS
Warning

Synonyms: (2-((Diphenylphosphino)methyl)-2-methylpropane-1,3-diyl)bis(diphenylphosphine), 1,1,1-Tris(diphenylphosphinomethyl)ethane

15-7870-5G 15-7870-1G
100208-358EA 334.03 USD
100208-358 100200-434
Triphos ≥97%
Triphos

Phosphine

Formula: C₄₁H₃₉P₃
MW: 624.684 g/mol
Melting Pt: 98…101 °C
MDL Number: MFCD00014084
CAS Number: 22031-12-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR