Tris(2,4-dimethylphenyl)phosphine ≥98%

Supplier: STREM CHEMICALS INC MS

15-7830-500MG 15-7830-2G
101418-716EA 120.31 USD
101418-716 101442-482
Tris(2,4-dimethylphenyl)phosphine ≥98%
Tris(2,4-dimethylphenyl)phosphine

Phosphine

Formula: C₂₄H₂₇P
MW: 346.4450 g/mol
Melting Pt: 157…158 °C
MDL Number: MFCD09039068
CAS Number: 49676-42-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR