Tris(3,5-dimethylphenyl)phosphine ≥98%

Supplier: STREM CHEMICALS INC MS
Warning

15-7820-500MG 15-7820-2G
100197-428EA 120.31 USD
100197-428 100211-030
Tris(3,5-dimethylphenyl)phosphine ≥98%
Tris(3,5-dimethylphenyl)phosphine

Phosphine

Formula: C₂₄H₂₇P
MW: 346.4450 g/mol
Melting Pt: 160…163 °C
MDL Number: MFCD03094579
CAS Number: 69227-47-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR