Tris(2,4-di-tert-butylphenyl)phosphite ≥98%

Supplier: STREM CHEMICALS INC MS

Synonyms: Tris(2,4-di-t-butylphenyl)phosphite, Phosphorous Acid Tris(2,4-di-tert-butylphenyl) Ester, Tris(2,4-di-tert-butylphenyl) phosphite

15-7720-250G 15-7720-1000G 15-7720-10G
100209-744EA 56.62 USD
100209-744 100198-684 100198-682
Tris(2,4-di-tert-butylphenyl)phosphite ≥98%
Tris(2,4-di-tert-butylphenyl)phosphite

Phosphine

Formula: C₄₂H₆₃O₃P
MW: 646.94 g/mol
Melting Pt: 181…184 °C
MDL Number: MFCD00072706
CAS Number: 31570-04-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR