TCEP-HCl (Tris(2-carboxyethyl)phosphine hydrochloride) ≥99%

Supplier: STREM CHEMICALS INC MS
Warning

Synonyms: 3,3',3''-Phosphinetriyltripropanoic acid hydrochloride, TCEP, Tris(2-carboxyethyl)phosphine, hydrochloride

15-7400-1G 15-7400-5G
100196-118EA 79.26 USD
100196-118 100203-014
TCEP-HCl (Tris(2-carboxyethyl)phosphine hydrochloride) ≥99%
TCEP-HCl (Tris(2-carboxyethyl)phosphine hydrochloride)

Phosphine, Phosphine, Water Soluble

Formula: C₉H₁₆ClO₆P
MW: 286.65 g/mol
MDL Number: MFCD00145469
CAS Number: 51805-45-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR