2,8,9-Tri-i-propyl-2,5,8,9-tetraaza-1-phosphabicyclo[3.3.3]undecane

Supplier: STREM CHEMICALS INC MS

15-7200-250MG 15-7200-1G
100198-676EA 84.92 USD
100198-676 100206-442
2,8,9-Tri-i-propyl-2,5,8,9-tetraaza-1-phosphabicyclo[3.3.3]undecane
2,8,9-Tri-i-propyl-2,5,8,9-tetraaza-1-phosphabicyclo[3.3.3]undecane

Phosphine

Density: 0,922
Flash Pt: 185 °F
MDL Number: MFCD03701530
CAS Number: 175845-21-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR