Tri(1-naphthyl)phosphine ≥98%

Supplier: STREM CHEMICALS INC MS

Synonyms: Tri(naphthalen-1-yl)phosphine

15-6610-5G 15-6610-1G
100196-108EA 276 USD
100196-108 100196-106
Tri(1-naphthyl)phosphine ≥98%
Tri(1-naphthyl)phosphine

Phosphine

Formula: C₃₀H₂₁P
MW: 412.46 g/mol
Melting Pt: 265…268 °C
MDL Number: MFCD00046400
CAS Number: 3411-48-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR