Trihexyl(tetradecyl)phosphonium chloride ≥93% (CYPHOS® IL 101)

Supplier: STREM CHEMICALS INC MS
Warning

15-6382-50G 15-6382-10G
100208-346EA 83.51 USD
100208-346 100208-344
Trihexyl(tetradecyl)phosphonium chloride ≥93% (CYPHOS® IL 101)
Trihexyl(tetradecyl)phosphonium chloride

CYPHOS IL, Ionic Liquids, Phosphine

Formula: C₃₂H₆₈ClP
MW: 519.31 g/mol
Density: 0,894
MDL Number: MFCD04112746
CAS Number: 258864-54-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR