Trihexyl(tetradecyl)phosphonium bromide ≥95% (CYPHOS® IL 102)

Supplier: STREM CHEMICALS INC MS
Warning

15-6378-10G 15-6378-50G
100208-340EA 43.88 USD
100208-340 100208-342
Trihexyl(tetradecyl)phosphonium bromide ≥95% (CYPHOS® IL 102)
Trihexyl(tetradecyl)phosphonium bromide

CYPHOS IL, Ionic Liquids, Phosphine

Formula: C₃₂H₆₈BrP
MW: 563.76 g/mol
Density: 0,952
MDL Number: MFCD04117697
CAS Number: 654057-97-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR