Trihexyl(tetradecyl)phosphonium bis(2,4,4-trimethylpentyl)phosphinate ≥95% (CYPHOS® IL 104)
Supplier: STREM CHEMICALS INC MS
|
Warning
|
15-6374-50G
15-6374-10G
100203-992EA
451.51
USD
100203-992
100203-990
Trihexyl(tetradecyl)phosphonium bis(2,4,4-trimethylpentyl)phosphinate ≥95% (CYPHOS® IL 104)
Trihexyl(tetradecyl)phosphonium bis(2,4,4-trimethylpentyl)phosphinate
CYPHOS IL, Ionic Liquids, Phosphine
Formula:
C₄₈H₁₀₂O₂P₂ MW: 773.27 g/mol Density: 0,887 |
MDL Number:
MFCD04117696 CAS Number: 465527-59-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...