Trihexyl(tetradecyl)phosphonium bis(trifluoromethylsulfonyl)imide ≥97% (CYPHOS® IL 109)
Supplier: STREM CHEMICALS INC MS
|
Warning
|
Synonyms:
Trihexyl(tetradecyl)phosphonium bis(trifluoromethanesulphonyl)amide
15-6370-50G
15-6370-10G
100207-354EA
404.8
USD
100207-354
100207-352
Trihexyl(tetradecyl)phosphonium bis(trifluoromethylsulfonyl)imide ≥97% (CYPHOS® IL 109)
Trihexyl(tetradecyl)phosphonium bis(trifluoromethylsulphonyl)imide
CYPHOS IL, Ionic Liquids, Phosphine
Formula:
C₃₄H₆₈F₆NO₄PS₂ |
MDL Number:
MFCD04117695 CAS Number: 460092-03-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...